Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-iodonitrobenzene 97%

Supplier: AMBEED, INC
Warning

A266032-10G A266032-1G A266032-250MG A266032-25G A266032-5G
76827-218EA 168 USD InStock
76827-218 76827-220 76827-222 76827-224 76827-226
3-Bromo-2-iodonitrobenzene 97%
3-Bromo-2-iodonitrobenzene
Precaución: for Research Use Only


Formula: C₆H₃BrINO₂
MDL Number: MFCD16660176
CAS Number: 32337-96-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty