Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(-)-Malic acid 98%

Supplier: AMBEED, INC
Warning

A266309-1000G A266309-100G A266309-25G A266309-500G
76827-228EA 44 USD InStock
76827-228 76827-230 76827-232 76827-234
L(-)-Malic acid 98%
L(-)-Malic acid
Precaución: for Research Use Only


Formula: C₄H₆O₅
MW: 134.09 g/mol
Boiling Pt: 322
MDL Number: MFCD00064213
CAS Number: 97-67-6
Merck Index: 13,05730
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty