Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dibromo-7-azaindole 95%

Supplier: AMBEED, INC
Warning

A270647-100MG A270647-1G A270647-250MG
76827-328EA 38 USD InStock
76827-328 76827-330 76827-332
3,4-Dibromo-7-azaindole 95%
3,4-Dibromo-7-azaindole
Precaución: for Research Use Only


Formula: C₇H₄Br₂N₂
MW: 275.92 g/mol
MDL Number: MFCD09880122
CAS Number: 1000340-33-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty