Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,2S)-2-(3,4-Difluorophenyl)cyclopropanamine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A272275-10G A272275-1G A272275-250MG A272275-25G A272275-5G
76827-360EA 7.43 USD InStock
76827-360 76827-362 76827-364 76827-366 76827-368 76827-370
(1R,2S)-2-(3,4-Difluorophenyl)cyclopropanamine hydrochloride 98%
(1R,2S)-2-(3,4-Difluorophenyl)cyclopropanamine hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₀ClF₂N
MDL Number: MFCD16621157
CAS Number: 1402222-66-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty