Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-(Trifluoromethyl)thiazol-2-yl)methanol 97%

Supplier: AMBEED, INC
Warning

A272385-100MG A272385-250MG A272385-5G
76827-374EA 39 USD InStock
76827-374 76827-376 76827-378
(4-(Trifluoromethyl)thiazol-2-yl)methanol 97%
(4-(Trifluoromethyl)thiazol-2-yl)methanol
Precaución: for Research Use Only


Formula: C₅H₄F₃NOS
MDL Number: MFCD09909578
CAS Number: 204319-69-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty