Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-6-trifluoromethylquinoline 95%

Supplier: AMBEED, INC
Warning

A284006-100MG A284006-10G A284006-1G A284006-250MG A284006-5G
76827-586EA 7 USD InStock
76827-586 76827-588 76827-590 76827-592 76827-594
4-Bromo-6-trifluoromethylquinoline 95%
4-Bromo-6-trifluoromethylquinoline
Precaución: for Research Use Only


Formula: C₁₀H₅BrF₃N
MW: 276.053 g/mol
MDL Number: MFCD11505113
CAS Number: 1070879-32-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty