Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-chloro-8,9-dihydro-5H-pyrimido[5,4-d]azepine-7(6H)-carboxylate 95%

Supplier: AMBEED, INC
Warning

A297810-100MG A297810-250MG A297810-50MG
76827-794EA 51 USD InStock
76827-794 76827-796 76827-798
tert-Butyl 4-chloro-8,9-dihydro-5H-pyrimido[5,4-d]azepine-7(6H)-carboxylate 95%
tert-Butyl 4-chloro-8,9-dihydro-5H-pyrimido[5,4-d]azepine-7(6H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₃H₁₈ClN₃O₂
MDL Number: MFCD11045433
CAS Number: 1057338-27-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty