Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2,3-difluoro-4-hydroxybenzoate ≥95%

Supplier: AMBEED, INC
Danger

A300500-100MG A300500-250MG
76827-852EA 41 USD InStock
76827-852 76827-854
Methyl 2,3-difluoro-4-hydroxybenzoate ≥95%
Methyl 2,3-difluoro-4-hydroxybenzoate
Precaución: for Research Use Only


Formula: C₈H₆F₂O₃
MDL Number: MFCD11042793
CAS Number: 219685-84-4
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty