Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methyl-3,4-dihydroisoquinolin-1(2H)-one 90%

Supplier: AMBEED, INC
Warning

A304088-100MG A304088-1G A304088-250MG
76827-912EA 16 USD InStock
76827-912 76827-914 76827-916
5-Methyl-3,4-dihydroisoquinolin-1(2H)-one 90%
5-Methyl-3,4-dihydroisoquinolin-1(2H)-one
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO
MDL Number: MFCD00890474
CAS Number: 129075-56-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty