Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-2-(hydroxymethyl)benzimidazole 98+%

Supplier: AMBEED, INC
Warning

A311821-100MG A311821-1G A311821-250MG A311821-5G
76828-008EA 5 USD InStock
76828-008 76828-010 76828-012 76828-014
5-Amino-2-(hydroxymethyl)benzimidazole 98+%
5-Amino-2-(hydroxymethyl)benzimidazole
Precaución: for Research Use Only


Formula: C₈H₉N₃O
MDL Number: MFCD00462584
CAS Number: 294656-36-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty