Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,2S,3R,3AR,3a1R,4R,6aR,7aS,11S,11aS,11bR)-1,2,3a,4,11-pentahydroxy-3,8,11a-trimethyl-1,2,3,3a,4,7,7a,11,11a,11b-decahydro-5H-3,3a1-(epoxymethano)dibenzo[de,g]chromene-5,10(6aH)-dione 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A313823-1MG A313823-10MG A313823-5MG
76828-058EA 41 USD InStock
76828-058 77683-931 77683-932
(1R,2S,3R,3AR,3a1R,4R,6aR,7aS,11S,11aS,11bR)-1,2,3a,4,11-pentahydroxy-3,8,11a-trimethyl-1,2,3,3a,4,7,7a,11,11a,11b-decahydro-5H-3,3a1-(epoxymethano)dibenzo[de,g]chromene-5,10(6aH)-dione 95%, Ambeed.Inc
(1R,2S,3R,3AR,3a1R,4R,6aR,7aS,11S,11aS,11bR)-1,2,3a,4,11-pentahydroxy-3,8,11a-trimethyl-1,2,3,3a,4,7,7a,11,11a,11b-decahydro-5H-3,3a1-(epoxymethano)dibenzo[de,g]chromene-5,10(6aH)-dione
Precaución: For research use only.


Formula: C₂₀H₂₆O₉
MDL Number: MFCD00221753
CAS Number: 21499-66-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty