Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,3R,4S,5R)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A319764-100G A319764-10G A319764-1G A319764-25G A319764-5G
76828-166EA 409 USD InStock
76828-166 76828-168 76828-170 76828-172 77683-949
(2R,3R,4S,5R)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol 95%, Ambeed.Inc
(2R,3R,4S,5R)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
Precaución: For research use only.


Formula: C₇H₁₄O₆
MDL Number: MFCD08692046
CAS Number: 93302-26-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty