Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Bromo-6-fluoroisoquinoline 97%

Supplier: AMBEED, INC
Warning

A322465-100MG A322465-1G A322465-250MG A322465-5G
76828-220EA 48 USD InStock
76828-220 76828-222 76828-224 76828-226
7-Bromo-6-fluoroisoquinoline 97%
7-Bromo-6-fluoroisoquinoline
Precaución: for Research Use Only


Formula: C₉H₅BrFN
MDL Number: MFCD11846300
CAS Number: 923022-40-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty