Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)porphyrin 98%

Supplier: AMBEED, INC
Warning

A325587-100MG A325587-1G A325587-250MG
76828-264EA 154 USD InStock
76828-264 76828-266 76828-268
5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)porphyrin 98%
5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)porphyrin
Precaución: for Research Use Only


Formula: C₄₄H₃₀N₄O₈
MW: 742.74 g/mol
MDL Number: MFCD02093497
CAS Number: 145764-54-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty