Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Odanacatib 98%

Supplier: AMBEED, INC
Warning

A328069-10MG A328069-1MG A328069-25MG A328069-2MG A328069-5MG
76828-306EA 171 USD InStock
76828-306 76828-308 76828-310 76828-312 76828-314
Odanacatib 98%
Odanacatib
Precaución: for Research Use Only


Formula: C₂₅H₂₇F₄N₃O₃S
MW: 525.56 g/mol
MDL Number: MFCD11042419
CAS Number: 603139-19-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty