Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyano-3,3-bis(methylthio)acrylamide 97%

Supplier: AMBEED, INC
Warning

A334591-100MG A334591-1G A334591-250MG A334591-5G
76828-388EA 39 USD InStock
76828-388 76828-390 76828-392 76828-394
2-Cyano-3,3-bis(methylthio)acrylamide 97%
2-Cyano-3,3-bis(methylthio)acrylamide
Precaución: for Research Use Only


Formula: C₆H₈N₂OS₂
MDL Number: MFCD00205587
CAS Number: 17823-69-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty