Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl phenyl carbonate 95%

Supplier: AMBEED, INC
Warning

A338983-10G A338983-1G A338983-25G A338983-5G
76828-444EA 13 USD InStock
76828-444 76828-446 76828-448 76828-450
Benzyl phenyl carbonate 95%
Benzyl phenyl carbonate
Precaución: for Research Use Only


Formula: C₁₄H₁₂O₃
MDL Number: MFCD07784341
CAS Number: 28170-07-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty