Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminopicolinamide 97%

Supplier: AMBEED, INC
Warning

A346437-1G A346437-250MG
76828-548EA 17 USD InStock
76828-548 76828-550
4-Aminopicolinamide 97%
4-Aminopicolinamide
Precaución: for Research Use Only


Formula: C₆H₇N₃O
MDL Number: MFCD08062892
CAS Number: 100137-47-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty