Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(N,N-dimethylaminopropyl)amine 98%

Supplier: AMBEED, INC
Danger

A350388-1G A350388-5G
76828-588EA 5 USD InStock
76828-588 76828-590
Tris(N,N-dimethylaminopropyl)amine 98%
Tris(N,N-dimethylaminopropyl)amine
Precaución: for Research Use Only


Formula: C₁₅H₃₆N₄
MDL Number: MFCD00190091
CAS Number: 33329-35-0
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty