Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-methyl-3-(trifluoromethyl)benzoate 95%

Supplier: AMBEED, INC
Warning

A351996-10G A351996-1G A351996-250MG A351996-5G
76828-592EA 86 USD InStock
76828-592 76828-594 76828-596 76828-598
Methyl 2-methyl-3-(trifluoromethyl)benzoate 95%
Methyl 2-methyl-3-(trifluoromethyl)benzoate
Precaución: For research use only.


Formula: C₁₀H₉F₃O₂
MW: 218.17 g/mol
MDL Number: MFCD14698066
CAS Number: 346603-63-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty