Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dibromo-2-methylpropane 95%

Supplier: AMBEED, INC
Warning

A357567-100G A357567-1G A357567-250MG A357567-25G A357567-5G
76828-696EA 294 USD InStock
76828-696 76828-698 76828-700 76828-702 76828-704
1,3-Dibromo-2-methylpropane 95%
1,3-Dibromo-2-methylpropane
Precaución: for Research Use Only


Formula: C₄H₈Br₂
MDL Number: MFCD18909351
CAS Number: 28148-04-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty