Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-(Neopentyloxy)phenyl)boronic acid 96%

Supplier: AMBEED, INC
Warning

A358717-1G A358717-250MG A358717-5G
76828-726EA 128 USD InStock
76828-726 76828-728 76828-730
(3-(Neopentyloxy)phenyl)boronic acid 96%
(3-(Neopentyloxy)phenyl)boronic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₇BO₃
MDL Number: MFCD18087686
CAS Number: 1236191-14-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty