Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methylselenocysteine 98%, Ambeed.Inc

Supplier: AMBEED, INC
Danger

A361358-100MG A361358-250MG A361358-50MG
76828-748EA 50 USD InStock
76828-748 76828-750 77684-061
Methylselenocysteine 98%, Ambeed.Inc
Methylselenocysteine
Precaución: For research use only.


Formula: C₄H₉NO₂Se
MW: 182.092 g/mol
MDL Number: MFCD00800565
CAS Number: 26046-90-2
UN: 3283
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty