Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Chlorophenyl)picolinic acid 95%

Supplier: AMBEED, INC
Warning

A362504-100MG A362504-1G A362504-250MG
76828-760EA 38 USD InStock
76828-760 76828-762 76828-764
4-(4-Chlorophenyl)picolinic acid 95%
4-(4-Chlorophenyl)picolinic acid
Precaución: for Research Use Only


Formula: C₁₂H₈ClNO₂
MW: 233.65 g/mol
MDL Number: MFCD18085942
CAS Number: 1258612-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty