Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-methoxy-4-pyrimidinamine 97%

Supplier: AMBEED, INC
Warning

A367660-10G A367660-1G A367660-250MG A367660-5G
76828-818EA 53 USD InStock
76828-818 76828-820 76828-822 76828-824
2-Chloro-5-methoxy-4-pyrimidinamine 97%
2-Chloro-5-methoxy-4-pyrimidinamine
Precaución: for Research Use Only


Formula: C₅H₆ClN₃O
MDL Number: MFCD17169913
CAS Number: 99979-77-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty