Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethylbenzenebutanoic acid 97%

Supplier: AMBEED, INC
Warning

A368010-250MG A368010-5G
76828-826EA 44 USD InStock
76828-826 76828-828
4-Ethylbenzenebutanoic acid 97%
4-Ethylbenzenebutanoic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₆O₂
MDL Number: MFCD01462080
CAS Number: 5467-53-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty