Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl isoquinoline-7-carboxylate 95%

Supplier: AMBEED, INC
Warning

A368067-100MG A368067-1G A368067-250MG
76828-830EA 25 USD InStock
76828-830 76828-832 76828-834
Methyl isoquinoline-7-carboxylate 95%
Methyl isoquinoline-7-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₉NO₂
MDL Number: MFCD11226962
CAS Number: 178262-31-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty