Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-nitrobenzoic acid 98%

Supplier: AMBEED, INC
Warning

A377368-1000G A377368-100G A377368-25G A377368-500G A377368-5G
76828-952EA 54 USD InStock
76828-952 76828-954 76828-956 76828-958 76828-960
4-Chloro-3-nitrobenzoic acid 98%
4-Chloro-3-nitrobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄ClNO₄
MDL Number: MFCD00007079
CAS Number: 96-99-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty