Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-(3-nitropyridin-2-yl)acetate 95%

Supplier: AMBEED, INC
Warning

A385451-100MG A385451-10G A385451-1G A385451-250MG A385451-5G
76829-070EA 45 USD InStock
76829-070 76829-072 76829-074 76829-076 76829-078
Ethyl 2-(3-nitropyridin-2-yl)acetate 95%
Ethyl 2-(3-nitropyridin-2-yl)acetate
Precaución: for Research Use Only


Formula: C₉H₁₀N₂O₄
MDL Number: MFCD11616147
CAS Number: 154078-83-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty