Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Boc-2-hydroxy-6-aza-spiro[3.4]octane 98%

Supplier: AMBEED, INC
Warning

A388661-100MG A388661-250MG
76829-092EA 28 USD InStock
76829-092 76829-094
6-Boc-2-hydroxy-6-aza-spiro[3.4]octane 98%
6-Boc-2-hydroxy-6-aza-spiro[3.4]octane
Precaución: for Research Use Only


Formula: C₁₂H₂₁NO₃
MDL Number: MFCD18073249
CAS Number: 1239319-91-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty