Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-methyl-1H-pyrrole-3-carbonitrile 95%

Supplier: AMBEED, INC
Warning

A396012-100MG A396012-1G A396012-250MG A396012-5G
76829-180EA 128 USD InStock
76829-180 76829-182 76829-184 76829-186
2-Amino-4-methyl-1H-pyrrole-3-carbonitrile 95%
2-Amino-4-methyl-1H-pyrrole-3-carbonitrile
Precaución: for Research Use Only


Formula: C₆H₇N₃
MDL Number: MFCD00128279
CAS Number: 59146-60-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty