Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

MK-1775 98%

Supplier: AMBEED, INC
Warning

A399066-100MG A399066-1MG A399066-5MG
76829-246EA 66 USD InStock
76829-246 76829-248 76829-250
MK-1775 98%
MK-1775
Precaución: for Research Use Only


Formula: C₂₇H₃₂N₈O₂
MDL Number: MFCD17215200
CAS Number: 955365-80-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty