Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Oxo-1,3-dihydroisobenzofuran-5-carbonitrile 97%

Supplier: AMBEED, INC
Warning

A400504-100MG A400504-1G A400504-250MG
76829-260EA 43 USD InStock
76829-260 76829-262 76829-264
3-Oxo-1,3-dihydroisobenzofuran-5-carbonitrile 97%
3-Oxo-1,3-dihydroisobenzofuran-5-carbonitrile
Precaución: for Research Use Only


Formula: C₉H₅NO₂
MDL Number: MFCD03428560
CAS Number: 89877-62-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty