Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-Fluoro-4'-nitroacetophenone 95%

Supplier: AMBEED, INC
Warning

A400939-1G A400939-250MG A400939-25G A400939-5G
76829-266EA 40 USD InStock
76829-266 76829-268 76829-270 76829-272
3'-Fluoro-4'-nitroacetophenone 95%
3'-Fluoro-4'-nitroacetophenone
Precaución: for Research Use Only


Formula: C₈H₆FNO₃
MDL Number: MFCD13194365
CAS Number: 72802-25-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty