Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium trifluoro(isoquinolin-4-yl)borate 98%

Supplier: AMBEED, INC
Warning

A414204-1G A414204-250MG A414204-5G
76829-366EA 65 USD InStock
76829-366 76829-368 76829-370
Potassium trifluoro(isoquinolin-4-yl)borate 98%
Potassium trifluoro(isoquinolin-4-yl)borate
Precaución: for Research Use Only


Formula: C₉H₆BF₃KN
MDL Number: MFCD08458186
CAS Number: 1111733-07-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty