Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(3,5-Dimethylphenyl)propanoic acid 95%

Supplier: AMBEED, INC
Warning

A414852-1G A414852-250MG
76829-376EA 66 USD InStock
76829-376 76829-378
3-(3,5-Dimethylphenyl)propanoic acid 95%
3-(3,5-Dimethylphenyl)propanoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD09055207
CAS Number: 42287-87-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty