Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Chlorophenyl)-4-oxobutanoic acid 97%

Supplier: AMBEED, INC
Warning

A415168-100MG A415168-1G A415168-250MG
76829-380EA 39 USD InStock
76829-380 76829-382 76829-384
4-(2-Chlorophenyl)-4-oxobutanoic acid 97%
4-(2-Chlorophenyl)-4-oxobutanoic acid
Precaución: for Research Use Only


Formula: C₁₀H₉ClO₃
MDL Number: MFCD03844205
CAS Number: 106263-50-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty