Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dioxo-3a,4,7,7a-tetrahydro-1H-4,7-methanoisoindol-2(3H)-yl 4-nitrobenzyl carbonate 97%

Supplier: AMBEED, INC
Warning

A425289-10G A425289-1G A425289-250MG A425289-25G A425289-5G
76829-466EA 34 USD InStock
76829-466 76829-468 76829-470 76829-472 76829-474
1,3-Dioxo-3a,4,7,7a-tetrahydro-1H-4,7-methanoisoindol-2(3H)-yl 4-nitrobenzyl carbonate 97%
1,3-Dioxo-3a,4,7,7a-tetrahydro-1H-4,7-methanoisoindol-2(3H)-yl 4-nitrobenzyl carbonate
Precaución: for Research Use Only


Formula: C₁₇H₁₄N₂O₇
MDL Number: MFCD18071454
CAS Number: 193269-82-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty