Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyano-2-fluorophenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A425334-100MG A425334-1G A425334-250MG A425334-5G
76829-476EA 15 USD InStock
76829-476 76829-478 76829-480 76829-482
4-Cyano-2-fluorophenylboronic acid pinacol ester 98%
4-Cyano-2-fluorophenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₅BFNO₂
MDL Number: MFCD09998162
CAS Number: 1035235-29-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty