Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromodibenzofuran 97%

Supplier: AMBEED, INC
Warning

A435139-100G A435139-10G A435139-1G A435139-25G A435139-500G A435139-5G
76829-574EA 87 USD InStock
76829-574 76829-576 76829-578 76829-580 76829-582 76829-584
4-Bromodibenzofuran 97%
4-Bromodibenzofuran
Precaución: for Research Use Only


Formula: C₁₂H₇BrO
MDL Number: MFCD00185685
CAS Number: 89827-45-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty