Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(((3aR,4S,6R,6aS)-6-Amino-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-yl)oxy)ethanol (2R,3R)-2,3-dihydroxysuccinate 98%

Supplier: AMBEED, INC
Warning

A444204-100G A444204-1G A444204-500G
76829-674EA 197 USD InStock
76829-674 76829-676 76829-678
2-(((3aR,4S,6R,6aS)-6-Amino-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-yl)oxy)ethanol (2R,3R)-2,3-dihydroxysuccinate 98%
2-(((3aR,4S,6R,6aS)-6-Amino-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-yl)oxy)ethanol (2R,3R)-2,3-dihydroxysuccinate
Precaución: for Research Use Only


Formula: C₁₄H₂₅NO₁₀
MW: 367.34 g/mol
MDL Number: MFCD22200635
CAS Number: 376608-65-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty