Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-bromo-6-chloropicolinate 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A451921-100MG A451921-1G A451921-250MG
76829-724EA 9 USD InStock
76829-724 76829-726 77684-236
Methyl 3-bromo-6-chloropicolinate 95%, Ambeed.Inc
Methyl 3-bromo-6-chloropicolinate
Precaución: For research use only.


Formula: C₇H₅BrClNO₂
MDL Number: MFCD11976777
CAS Number: 1214328-96-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty