Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((tert-Butoxycarbonyl)amino)ethyl methacrylate 95%

Supplier: AMBEED, INC
Warning

A452883-100MG A452883-1G A452883-250MG
76829-728EA 13 USD InStock
76829-728 76829-730 76829-732
2-((tert-Butoxycarbonyl)amino)ethyl methacrylate 95%
2-((tert-Butoxycarbonyl)amino)ethyl methacrylate
Precaución: for Research Use Only


Formula: C₁₁H₁₉NO₄
MDL Number: MFCD23140962
CAS Number: 89743-52-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty