Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

γ-Secretase inhibitor XX 95%

Supplier: AMBEED, INC
Warning

A457173-10MG A457173-1MG A457173-25MG A457173-2MG A457173-5MG
76829-776EA 229 USD InStock
76829-776 76829-778 76829-780 76829-782 76829-784
γ-Secretase inhibitor XX 95%
gamma-Secretase inhibitor XX
Precaución: for Research Use Only


Formula: C₂₆H₂₃F₂N₃O₃
MW: 463.48 g/mol
MDL Number: MFCD12828734
CAS Number: 209984-56-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty