Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Chlorobutyl)pyridine hydrochloride 95%

Supplier: AMBEED, INC
Danger

A461253-10G A461253-1G A461253-5G
76829-828EA 184 USD InStock
76829-828 76829-830 76829-832
4-(4-Chlorobutyl)pyridine hydrochloride 95%
4-(4-Chlorobutyl)pyridine hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₃Cl₂N
MDL Number: MFCD09952022
CAS Number: 149463-65-0
UN: 3261
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty