Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GW-0742 95%

Supplier: AMBEED, INC
Warning

A469160-10MG A469160-50MG
76829-896EA 5 USD InStock
76829-896 76829-898
GW-0742 95%
GW-0742
Precaución: for Research Use Only


Formula: C₂₁H₁₇F₄NO₃S₂
MDL Number: MFCD07369423
CAS Number: 317318-84-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty