Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluorophenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A479266-1G A479266-250MG A479266-25G A479266-5G
76829-986EA 36 USD InStock
76829-986 76829-988 76829-990 76829-992
3,5-Difluorophenylboronic acid pinacol ester 98%
3,5-Difluorophenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₁₅BF₂O₂
MDL Number: MFCD11504963
CAS Number: 863868-36-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty