Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-nitrophenyl methyl sulfone 95%

Supplier: AMBEED, INC
Warning

A493536-100G A493536-10G A493536-1G A493536-5G
76830-092EA 103 USD InStock
76830-092 76830-094 76830-096 76830-098 76830-100
4-Chloro-3-nitrophenyl methyl sulfone 95%
4-Chloro-3-nitrophenyl methyl sulfone
Precaución: for Research Use Only


Formula: C₇H₆ClNO₄S
MDL Number: MFCD00039754
CAS Number: 97-07-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty