Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-L-tryptophan 95%

Supplier: AMBEED, INC
Warning

A499754-100MG A499754-1G A499754-250MG
76830-170EA 27 USD InStock
76830-170 76830-172 76830-174
6-Bromo-L-tryptophan 95%
6-Bromo-L-tryptophan
Precaución: for Research Use Only


Formula: C₁₁H₁₁BrN₂O₂
MW: 283.12 g/mol
MDL Number: MFCD17214433
CAS Number: 52448-17-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty