Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Magnesium 2-aminoacetate 98%

Supplier: AMBEED, INC
Warning

A503602-100G A503602-25G
76830-212EA 22 USD InStock
76830-212 76830-214
Magnesium 2-aminoacetate 98%
Magnesium 2-aminoacetate
Precaución: for Research Use Only


Formula: C₄H₈MgN₂O₄
MDL Number: MFCD00274170
CAS Number: 14783-68-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty